Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutyl(Trimethoxy)Silane

Catalog No. 44406525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-406-525ML 25 mL
44-406-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 44-406-525ML Supplier Sigma Aldrich Supplier No. 44406525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobutyl(trimethoxy)silane-modified substrates were used for the preparation of gold films by absorption and adsorption.{87} It may be used as silane precursors for the synthesis of xerogels.{88}

Specifications

CAS 18395-30-7
Density 0.93 g/mL at 25°C
Boiling Point 154°C
Molecular Formula C7H18O3Si
Refractive Index n20/D 1.396
Linear Formula (CH3)2CHCH2Si(OCH3)3
MDL Number MFCD00051589
Quantity 25 mL
Molecular Weight (g/mol) 178.3 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.