accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CSiClClCH3CH3H3C

CAS RN 18395-90-9

IUPAC Name: di-tert-butyldichlorosilane
Synonyms: di-tert-butyldichlorosilane
Molecular Weight (g/mol): 213.22
Molecular Formula: C8H18Cl2Si
InChi Key: PDYPRPVKBUOHDH-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](Cl)(Cl)C(C)(C)C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)