Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyldichlorosilane 95.0+%, TCI America™
SDP

Catalog No. D24695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24695G Supplier TCI America Supplier No. D24695G

Chemical Identifiers

CAS 18395-90-9
Molecular Formula C8H18Cl2Si
Molecular Weight (g/mol) 213.217
MDL Number MFCD00008796
InChI Key PDYPRPVKBUOHDH-UHFFFAOYSA-N
PubChem CID 87624
IUPAC Name ditert-butyl(dichloro)silane
SMILES CC(C)(C)[Si](C(C)(C)C)(Cl)Cl

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 191°C
Quantity 5 g
UN Number 2987
Formula Weight 213.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-butyldichlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.