accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1843-05-6

IUPAC Name: 2-benzoyl-5-(octyloxy)phenol
Synonyms: benzophenone 12 uvinul M 408 sanduvor 3035 seikalizer E chimassorb 81 rhodialux P benzon OO uvinul 408 spectra-sorb UV 531 UV 1 (ultroviolet absorber)
Molecular Weight (g/mol): 326.44
Molecular Formula: C21H26O3
InChi Key: QUAMTGJKVDWJEQ-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)