Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-n-octyloxybenzophenone, 99%, Thermo Scientific™

Catalog No. AAL1175022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1175022 100 g
AAL1175014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1175022 Supplier Thermo Scientific Chemicals Supplier No. L1175022

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1843-05-6
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.436
MDL Number MFCD00027327
InChI Key QUAMTGJKVDWJEQ-UHFFFAOYSA-N
Synonym octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130
PubChem CID 15797
IUPAC Name (2-hydroxy-4-octoxyphenyl)-phenylmethanone
SMILES CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 46°C to 49°C
Quantity 100 g
Beilstein 1915198
Merck Index 14,6742
Formula Weight 326.44
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-4-n-octyloxybenzophenone
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 217-421-2
RTECSNumber DJ1595000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.