accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18437-66-6

IUPAC Name: tert-butyl N-(4-chlorophenyl)carbamate
Synonyms: tert-butyl N-(4-chlorophenyl)carbamate
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: VFEHOBXXLPHSOI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)