Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-chloroaniline, 98%, Thermo Scientific™

Catalog No. AC425890250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC425890250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425890250 Supplier Thermo Scientific Chemicals Supplier No. 425890250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 18437-66-6
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.688
MDL Number MFCD00816772
InChI Key VFEHOBXXLPHSOI-UHFFFAOYSA-N
Synonym tert-butyl 4-chlorophenyl carbamate, tert-butyl n-4-chlorophenyl carbamate, tert-butyl 4-chlorophenylcarbamate, 4-chloro-n-boc aniline, n-tert-butoxycarbonyl-4-chloroaniline, tert-butyl4-chlorophenylcarbamate, n-boc-4-chloroaniline, 4-chloro-n-boc-aniline, maybridge1_008229
PubChem CID 546372
IUPAC Name tert-butyl N-(4-chlorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)Cl

Specifications

Melting Point 102°C to 106°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
NMR Consistent with structure
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula ClC6H4NHCOOC(CH3)3
Quantity 25 g
Formula Weight 227.69
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-4-Chloroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.