accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18438-38-5

IUPAC Name: 2-(methylsulfanyl)pyridine
Synonyms: 2-methylthiopyridine
Molecular Weight (g/mol): 125.19
Molecular Formula: C6H7NS
InChi Key: VLQBSKLZRSUMTJ-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results