Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)pyridine 98.0+%, TCI America™
SDP

Catalog No. M184925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M184925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M184925G Supplier TCI America Supplier No. M184925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18438-38-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.189
MDL Number MFCD01691593
InChI Key VLQBSKLZRSUMTJ-UHFFFAOYSA-N
Synonym Methyl 2-Pyridyl Sulfide
PubChem CID 29076
IUPAC Name 2-methylsulfanylpyridine
SMILES CSC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 125.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)pyridine
RTECSNumber UT6207000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.