accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18439-24-2

IUPAC Name: trilithium(1+) (3R)-3-hydroxy-2,2-dimethyl-3-({2-[(2-sulfanylethyl)carbamoyl]ethyl}carbamoyl)propyl ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-(hydrogen phosphonatooxy)-4-hydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: trilithium(1+) (3R)-3-hydroxy-2,2-dimethyl-3-({2-[(2-sulfanylethyl)carbamoyl]ethyl}carbamoyl)propyl {[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3-(hydrogen phosphonatooxy)-4-hydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 785.33
Molecular Formula: C21H33Li3N7O16P3S
InChi Key: QSCBPHBAFBVXRK-FFKZSOHASA-K
SMILES: [Li+].[Li+].[Li+].CC(C)(COP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)([O-])=O)N1C=NC2=C(N)N=CN=C12)[C@@H](O)C(=O)NCCC(=O)NCCS
Physical Form 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)