Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coenzyme A Trilithium Salt Dihydrate, MP Biomedicals™

Catalog No. MP210049391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210049391 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210049391 Supplier MP Biomedicals, Inc Supplier No. 0210049391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Facilitates removal of lipid peroxides and promotes repair of plasma membranes. Coenzyme A Trilithium Salt Dihydrate, MP Biomedicals is an essential cofactor in enzymatic acetyl transfer reactions. It increases mobilization of fatty acids and activates ph

The principal biologically active forms of pantothenic acid are coenzyme A (CoA) and acyl carrier protein (ACP). In CoA, the business center of the molecule is the pantothenic acid metabolite 4'-phosphopantetheine.

  • Coenzyme A is comprised of 4'-phosphopantetheine linked by an anhydride bond to the nucloetide adenosine 5'-monophosphate
  • 4'-Phosphopantetheine itself is comprised of pantothenic acid linked at one end, via an amide bond, to beta-mercaptoethylamine, derived from L-cysteine, and at the other end to a phosphate group
  • The sulfhydryl group of 4'-phosphopantetheine, which is the business end of the coenzyme, forms thioesters with acyl groups producing acyl-CoA derivatives, including acetyl-CoA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Molecular Weight (g/mol) 785.329
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
Synonym coenzyme a, trilithium salt
PubChem CID 53442190
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O

Specifications

Quantity 1 g
Solubility Information Soluble in water (50mg/mL, clear, colorless to faint yellow solution)
Formula Weight 821.4
Percent Purity ≥96%
Physical Form Crystalline Powder
Recommended Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.