accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COBr

CAS RN 18469-37-9

IUPAC Name: 4-bromo-N,N-dimethylbenzamide
Synonyms: 4-bromo-N,N-dimethylbenzamide
Molecular Weight (g/mol): 228.09
Molecular Formula: C9H10BrNO
InChi Key: BRRVYBRXLUEEJP-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results