accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1847-24-1

IUPAC Name: sodium (2S,5R,6R)-6-[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: flucloxacillin sodium flucloxacillin sodium salt
Molecular Weight (g/mol): 475.85
Molecular Formula: C19H16ClFN3NaO5S
InChi Key: OTEANHMVDHZOPB-SLINCCQESA-M
SMILES: [Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=C(F)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)