Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Flucloxacillin, sodium salt, 95%, Thermo Scientific Chemicals

Catalog No. AC456470010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456470010 Supplier Thermo Scientific Chemicals Supplier No. 456470010
  • $198.00
    $157.65 / Each of 1

    Save $40.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1847-24-1
Molecular Formula C19H16ClFN3NaO5S
Molecular Weight (g/mol) 475.85
MDL Number MFCD01682108
InChI Key OTEANHMVDHZOPB-SLINCCQESA-M
PubChem CID 101643208
ChEBI CHEBI:31615
SMILES [Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

Type Flucloxacillin Sodium Salt
Melting Point 176.0°C to 178.0°C
Infrared Spectrum Authentic
Loss on Drying 4.5% max.
Specific Rotation +158° to +168° (20°C, 589nm) (c=1, water) On dry substance
Quantity 1 g
Flash Point >110°C
Boiling Point 677.0°C
IUPAC Name sodium;N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptan-6-yl]-3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboximidate
Formula Weight 475.86
Percent Purity 98%
Chemical Name or Material Flucloxacillin, sodium salt
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. In case of inadequate ventilation wear respiratory protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.