accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1849-01-0

IUPAC Name: 1-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-methyl-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: PYEHNKXDXBNHQQ-UHFFFAOYSA-N
SMILES: CN1C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results