Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-benzimidazolinone, 98%

Catalog No. AC276500050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276500050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276500050 Supplier Thermo Scientific Chemicals Supplier No. 276500050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1849-01-0 | C8H8N2O | 148.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1849-01-0
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
MDL Number MFCD00142654
InChI Key PYEHNKXDXBNHQQ-UHFFFAOYSA-N
Synonym 1-methyl-2-benzimidazolinone, 1-methyl-1h-benzo d imidazol-2 3h-one, 1-methyl-1,3-dihydro-2h-benzimidazol-2-one, 1,3-dihydro-1-methyl-2h-benzimidazol-2-one, 2h-benzimidazol-2-one, 1,3-dihydro-1-methyl, 1-methyl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-methyl-3h-1,3-benzodiazol-2-one, n-methylbenzimidazolone, 1-methylbenzimidazolone, n-methylbenzimidazolinone
PubChem CID 590540
IUPAC Name 3-methyl-1H-benzimidazol-2-one
SMILES CN1C(=O)NC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 192°C to 196°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, IV, 276
Formula Weight 148.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-2-benzimidazolinone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.