accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18498-01-6

IUPAC Name: [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichlorocobaltbis(ylium)
Synonyms: dichlorocobaltbis(ylium); diphos
Molecular Weight (g/mol): 528.26
Molecular Formula: C26H24Cl2CoP2
InChi Key: RWOAVAAHCHFKHE-UHFFFAOYSA-L
SMILES: Cl[Co++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results