accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOON

CAS RN 1851-09-8

IUPAC Name: 2-(4-chlorobenzenesulfonyl)acetonitrile
Synonyms: 2-(4-chlorobenzenesulfonyl)acetonitrile
Molecular Weight (g/mol): 215.65
Molecular Formula: C8H6ClNO2S
InChi Key: HAQGVGPNKGGSMK-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)CC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results