accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 1852-04-6

IUPAC Name: undecanedioic acid
Synonyms: undecanedioic acid
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: LWBHHRRTOZQPDM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)