Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanedioic Acid 97%, Thermo Scientific™

Catalog No. AC176160050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176160050 Supplier Thermo Scientific Chemicals Supplier No. 176160050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1852-04-6
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
InChI Key LWBHHRRTOZQPDM-UHFFFAOYSA-N
Synonym 1,9-nonanedicarboxylic acid, 1,11-undecanedioic acid, undecanedionic acid, 1,9-nonanedicarboxylicacid, undecandioic acid, hendecanedioic acid, unii-7059gfk8nv, nonamethylenedicarboxylic acid, hendecanedioate, undecanedioate
PubChem CID 15816
ChEBI CHEBI:73713
IUPAC Name undecanedioic acid
SMILES OC(=O)CCCCCCCCCC(O)=O

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Linear Formula HO2C(CH2)9CO2H
Quantity 5 g
Beilstein 02, 727
Formula Weight 216.28
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Undecanedioic acid, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.