accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1852-16-0

IUPAC Name: N-(butoxymethyl)prop-2-enamide
Synonyms: N-(butoxymethyl)acrylamide
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: UTSYWKJYFPPRAP-UHFFFAOYSA-N
SMILES: CCCCOCNC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results