Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Butoxymethyl)acrylamide (stabilized with MEHQ) 96.0+%, TCI America™
SDP

Catalog No. B1060500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1060500ML 500 mL
B106025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1060500ML Supplier TCI America Supplier No. B1060500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1852-16-0
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00059417
InChI Key UTSYWKJYFPPRAP-UHFFFAOYSA-N
Synonym n-butoxymethyl acrylamide, 2-propenamide, n-butoxymethyl, n-butoxymethylacrylamide, n-butoxymethylakrylamid, n-butoxymethyl-2-propenamide, acrylamide, n-butoxymethyl, n-n-butoxymethyl acrylamide, unii-z89g4o958a, n-butoxymethyl prop-2-enamide, n-butoxymethylakrylamid czech
PubChem CID 15817
IUPAC Name N-(butoxymethyl)prop-2-enamide
SMILES CCCCOCNC(=O)C=C

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 157.21
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material N-(Butoxymethyl)acrylamide (stabilized with MEHQ)
EINECSNumber (2)-1021&(2)-1070
RTECSNumber AS3450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.