accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18527-21-4

IUPAC Name: 2-[(4-fluorophenyl)sulfanyl]acetonitrile
Synonyms: 2-[(4-fluorophenyl)sulfanyl]acetonitrile
Molecular Weight (g/mol): 167.2
Molecular Formula: C8H6FNS
InChi Key: BJFLSPRQCMQITC-UHFFFAOYSA-N
SMILES: FC1=CC=C(SCC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results