Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Fluorophenylthio)acetonitrile, 97%, Thermo Scientific™

Catalog No. AAB2052603
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2052603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2052603 Supplier Thermo Scientific Chemicals Supplier No. B2052603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18527-21-4
Molecular Formula C8H6FNS
MDL Number MFCD00179342
Odor Unpleasant
Quantity 1g
Synonym 4-fluorophenylthio acetonitrile, 2-4-fluorophenyl thio acetonitrile, 2-4-fluorophenyl sulfanyl acetonitrile, acetonitrile,2-4-fluorophenyl thio, 2-4-fluorophenylthio ethanenitrile, acmc-1bnjb, maybridge1_004450, 2-4-fluorophenylthio acetonitrile
InChI Key BJFLSPRQCMQITC-UHFFFAOYSA-N
SMILES FC1=CC=C(SCC#N)C=C1
IUPAC Name 2-[(4-fluorophenyl)sulfanyl]acetonitrile
Molecular Weight (g/mol) 167.20
PubChem CID 581375
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material (4-Fluorophenylthio)acetonitrile
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.