accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiOCH3OCH3OCH3

CAS RN 18536-91-9

IUPAC Name: dodecyltriethoxysilane
Synonyms: dodecyltriethoxysilane
Molecular Weight (g/mol): 332.6
Molecular Formula: C18H40O3Si
InChi Key: YGUFXEJWPRRAEK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[Si](OCC)(OCC)OCC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results