Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dodecyltriethoxysilane

Catalog No. 4423725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-237-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-237-25ML Supplier Sigma Aldrich Supplier No. 4423725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reaction between dodecene and triethoxysilane yields dodecyltriethoxysilane. 1Dodecyltriethoxysilane can be used in the surface treatment of calcite crystals for the determination of the effect of wettability with the incorporation of nanoparticles. It may be electrodeposited with tetraethoxysilane, which can be used as an anti-corrosive coating on mild steel.

Specifications

CAS 18536-91-9
Density 0.875 g/mL (at 20°C)
Molecular Formula C18H40O3Si
Linear Formula C18H40O3Si
Quantity 25 mL
MDL Number MFCD00048437
Synonym 1-(Triethoxysilyl)dodecane; Dodecyltriethoxysilane; Triethoxydodecylsilane
Molecular Weight (g/mol) 332.59
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.