accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOONHN(R)

CAS RN 185379-39-9

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-2-yl)propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid
Molecular Weight (g/mol): 388.42
Molecular Formula: C23H20N2O4
InChi Key: DXIVJCDZOMUITR-OAQYLSRUSA-N
SMILES: OC(=O)[C@@H](CC1=NC=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)