accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185416-17-5

CH3CH3OCH3MgBr

CAS RN 185416-17-5

IUPAC Name: bromo(4-methoxy-3,5-dimethylphenyl)magnesium
Synonyms: bromo(4-methoxy-3,5-dimethylphenyl)magnesium
Molecular Weight (g/mol): 239.4
Molecular Formula: C9H11BrMgO
InChi Key: KIYSUNNVTFBQIT-UHFFFAOYSA-M
SMILES: COC1=C(C)C=C([Mg]Br)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results