accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOOHOHOHOHOOOONOONHOCH3HOHOHO

CAS RN 1858224-04-0

IUPAC Name: 5-acetamido-2-{[5-acetamido-3,4-dihydroxy-6-(4-nitrophenoxy)oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 5-acetamido-2-{[5-acetamido-3,4-dihydroxy-6-(4-nitrophenoxy)oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 633.56
Molecular Formula: C25H35N3O16
InChi Key: LRTAJFSVNJSJQO-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(COC2(CC(O)C(NC(C)=O)C(O2)C(O)C(O)CO)C(O)=O)OC1OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results