Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neu5Ac alpha(2-6)GalNAc-alpha-pNP 98.0+%, TCI America™
SDP

Catalog No. N08905MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08905MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08905MG Supplier TCI America Supplier No. N08905MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1858224-04-0
Molecular Formula C25H35N3O16
Molecular Weight (g/mol) 633.56
MDL Number MFCD20265599
InChI Key LRTAJFSVNJSJQO-UHFFFAOYNA-N
PubChem CID 121235357
IUPAC Name 5-acetamido-2-{[5-acetamido-3,4-dihydroxy-6-(4-nitrophenoxy)oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
SMILES CC(=O)NC1C(O)C(O)C(COC2(CC(O)C(NC(C)=O)C(O2)C(O)C(O)CO)C(O)=O)OC1OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 mg
Formula Weight 633.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac alpha(2-6)GalNAc-alpha-pNP
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.