accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOOOONNNHOOHOHHOOHOH

CAS RN 1858224-15-3

IUPAC Name: N-(2-{[2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Synonyms: N-(2-{[2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Molecular Weight (g/mol): 452.42
Molecular Formula: C16H28N4O11
InChi Key: BFBRNUJPGYGVRQ-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(O)C(CO)OC1OCCN=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results