Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GlcNAc beta(1-2)Man-alpha-ethylazide 98.0+%, TCI America™
SDP

Catalog No. G0337100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0337100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0337100MG Supplier TCI America Supplier No. G0337100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1858224-15-3
Molecular Formula C16H28N4O11
Molecular Weight (g/mol) 452.42
MDL Number MFCD15072164
InChI Key BFBRNUJPGYGVRQ-UHFFFAOYNA-N
PubChem CID 121234054
IUPAC Name N-(2-{[2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
SMILES CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(O)C(CO)OC1OCCN=[N+]=[N-]

Specifications

Color White
Quantity 100 mg
Formula Weight 452.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-2)Man-alpha-ethylazide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.