accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONHN

CAS RN 1858240-50-2

IUPAC Name: 2-(6-methyl-3-oxo-3,4-dihydroquinoxalin-2-yl)-2-phenylacetonitrile
Synonyms: 2-(6-methyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
Molecular Weight (g/mol): 275.31
Molecular Formula: C17H13N3O
InChi Key: VBHNCMAFPOPJJC-UHFFFAOYNA-N
SMILES: CC1=CC=C2N=C(C(C#N)C3=CC=CC=C3)C(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results