accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNFFFN

CAS RN 1858240-54-6

IUPAC Name: 4-phenyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
Synonyms: 4-phenyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol): 330.33
Molecular Formula: C17H9F3N2S
InChi Key: XUFKMNGEUDCIIV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results