Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-4-phenyl-2-[4-(trifluoromethyl)phenyl]thiazole, 97%, Thermo Scientific™

Catalog No. AAH5859906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5859906 5 g
AAH5859903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5859906 Supplier Thermo Scientific Chemicals Supplier No. H5859906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858240-54-6
Molecular Formula C17H9F3N2S
Molecular Weight (g/mol) 330.33
MDL Number MFCD22682993
InChI Key XUFKMNGEUDCIIV-UHFFFAOYSA-N
Synonym 4-phenyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-4-trifluoromethyl phenyl thiazole
PubChem CID 98000107
IUPAC Name 4-phenyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
SMILES FC(F)(F)C1=CC=C(C=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 330.33
Percent Purity 97%
Chemical Name or Material 5-Cyano-4-phenyl-2-[4-(trifluoromethyl)phenyl]thiazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.