accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSOONHOCH3

CAS RN 1858240-55-7

IUPAC Name: N-{4-[(4-methylpiperidin-2-yl)sulfonyl]phenyl}acetamide
Synonyms: N-[4-(4-methylpiperidin-2-ylsulfonyl)phenyl]acetamide
Molecular Weight (g/mol): 296.39
Molecular Formula: C14H20N2O3S
InChi Key: LUPVXSOGPVCCJN-UHFFFAOYNA-N
SMILES: CC1CCNC(C1)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results