accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1858241-81-2

IUPAC Name: 3-(4-bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
Molecular Weight (g/mol): 314.19
Molecular Formula: C15H12BrN3
InChi Key: TYXCTRHZBUPEHB-UHFFFAOYSA-N
SMILES: CC1=NN=C(N1C1=CC=CC=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results