Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole, 97%, Thermo Scientific™

Catalog No. AAH60327ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60327ME 500 mg
AAH60327MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60327ME Supplier Thermo Scientific Chemicals Supplier No. H60327ME

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858241-81-2
Molecular Formula C15H12BrN3
Molecular Weight (g/mol) 314.186
MDL Number MFCD22683118
InChI Key TYXCTRHZBUPEHB-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
PubChem CID 98000572
IUPAC Name 3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br

Specifications

Quantity 500 mg
Formula Weight 314.18
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.