CAS RN 1858241-97-0
IUPAC Name:
2-(3-methylpyridin-2-yl)-4-phenyl-1,3-thiazole-5-carbonitrile
Synonyms:
2-(3-methylpyridin-2-yl)-4-phenyl-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol):
277.35
Molecular Formula:
C16H11N3S
InChi Key:
GNCHRPZBZFYZAD-UHFFFAOYSA-N
SMILES:
CC1=C(N=CC=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Cyano-2-(3-methyl-2-pyridyl)-4-phenylthiazole, 97%, Thermo Scientific™
CAS: 1858241-97-0 Molecular Formula: C16H11N3S Molecular Weight (g/mol): 277.345 MDL Number: MFCD22682957 InChI Key: GNCHRPZBZFYZAD-UHFFFAOYSA-N Synonym: 2-3-methylpyridin-2-yl-4-phenyl-1,3-thiazole-5-carbonitrile,5-cyano-2-3-methyl-2-pyridyl-4-phenylthiazole PubChem CID: 98000062 IUPAC Name: 2-(3-methylpyridin-2-yl)-4-phenyl-1,3-thiazole-5-carbonitrile SMILES: CC1=C(N=CC=C1)C2=NC(=C(S2)C#N)C3=CC=CC=C3