accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSOOCH3NH2

CAS RN 1858250-46-0

IUPAC Name: ethyl 4-[4-(acetyloxy)phenyl]-2-aminothiophene-3-carboxylate
Synonyms: ethyl 4-[4-(acetyloxy)phenyl]-2-aminothiophene-3-carboxylate
Molecular Weight (g/mol): 305.35
Molecular Formula: C15H15NO4S
InChi Key: RAXUJVOBTPWILV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(OC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results