accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1858252-13-7

IUPAC Name: 1-(4-phenyl-1,3-thiazol-2-yl)piperidine-4-carbonitrile
Synonyms: 1-(4-phenyl-1,3-thiazol-2-yl)piperidine-4-carbonitrile
Molecular Weight (g/mol): 269.37
Molecular Formula: C15H15N3S
InChi Key: KTSVMKZSGOBSDS-UHFFFAOYSA-N
SMILES: N#CC1CCN(CC1)C1=NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results