accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNN

CAS RN 1858252-21-7

IUPAC Name: 4-phenyl-2-(pyridin-3-yl)-1,3-thiazole-5-carbonitrile
Synonyms: 4-phenyl-2-(pyridin-3-yl)-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol): 263.32
Molecular Formula: C15H9N3S
InChi Key: QBABKNRVLIDXCF-UHFFFAOYSA-N
SMILES: N#CC1=C(N=C(S1)C1=CN=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results