accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1858255-54-5

IUPAC Name: 3-(4-bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 374.24
Molecular Formula: C17H16BrN3O2
InChi Key: SPFFEOOTWDLJSH-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=C1)N1C(C)=NN=C1C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results