CAS RN 1858256-22-0
IUPAC Name:
ethyl 4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylate
Synonyms:
ethyl 4-[(2,5-dioxopyrrol-1-yl)methyl]-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol):
343.36
Molecular Formula:
C16H13N3O4S
InChi Key:
LSOCKOVBJWMGKX-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(CN2C(=O)C=CC2=O)N=C(S1)C1=CC=CC=N1
1
–
1
of
1
results
1
Ethyl 4-(maleimidomethyl)-2-(2-pyridyl)thiazole-5-carboxylate, 97%, Thermo Scientific™
CAS: 1858256-22-0 Molecular Formula: C16H13N3O4S Molecular Weight (g/mol): 343.357 MDL Number: MFCD22988977 InChI Key: LSOCKOVBJWMGKX-UHFFFAOYSA-N Synonym: ethyl 4-2,5-dioxopyrrol-1-yl methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate,ethyl 4-maleimidomethyl-2-2-pyridyl thiazole-5-carboxylate PubChem CID: 98000538 IUPAC Name: ethyl 4-[(2,5-dioxopyrrol-1-yl)methyl]-2-pyridin-2-yl-1,3-thiazole-5-carboxylate SMILES: CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)CN3C(=O)C=CC3=O