accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOHNH2OOCH3(E/Z)

CAS RN 1858264-20-6

IUPAC Name: 2-(acetyloxy)-5-(N'-hydroxycarbamimidoyl)phenyl acetate
Synonyms: 2-(acetyloxy)-5-(N'-hydroxycarbamimidoyl)phenyl acetate
Molecular Weight (g/mol): 252.23
Molecular Formula: C11H12N2O5
InChi Key: DUQSSXZZHVNOOD-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(OC(C)=O)C=C(C=C1)C(N)=NO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results