accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNNH2NOS(E/Z)(E/Z)

CAS RN 1858264-30-8

IUPAC Name: 1-[2-(3-methyl-1,2-dihydropyridin-2-ylidene)-4-phenyl-2,5-dihydro-1,3-thiazol-5-ylidene]-1-nitrosomethanamine
Synonyms: 1-[2-(3-methyl-1H-pyridin-2-ylidene)-4-phenyl-1,3-thiazol-5-ylidene]-1-nitrosomethanamine
Molecular Weight (g/mol): 310.38
Molecular Formula: C16H14N4OS
InChi Key: PGZZXMLURFEAJX-UHFFFAOYSA-N
SMILES: CC1=CC=CNC1=C1SC(=C(N)N=O)C(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results