accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNSH2NNO(E/Z)(E/Z)

CAS RN 1858264-54-6

IUPAC Name: 1-[2-(1,2-dihydropyrazin-2-ylidene)-4-phenyl-2,5-dihydro-1,3-thiazol-5-ylidene]-1-nitrosomethanamine
Synonyms: 1-nitroso-1-[4-phenyl-2-(1H-pyrazin-2-ylidene)-1,3-thiazol-5-ylidene]methanamine
Molecular Weight (g/mol): 297.34
Molecular Formula: C14H11N5OS
InChi Key: JCKDRHGFBLHZES-UHFFFAOYSA-N
SMILES: NC(N=O)=C1SC(N=C1C1=CC=CC=C1)=C1NC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results