accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClP

CAS RN 18583-55-6

IUPAC Name: [(2-chlorophenyl)methyl]triphenylphosphanium chloride
Synonyms: [(2-chlorophenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 423.32
Molecular Formula: C25H21Cl2P
InChi Key: VRAAQIWOSHYDHE-UHFFFAOYSA-M
SMILES: [Cl-].ClC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results