Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Chlorobenzyl)triphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. C175925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C175925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C175925G Supplier TCI America Supplier No. C175925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18583-55-6
Molecular Formula C25H21Cl2P
Molecular Weight (g/mol) 423.32
MDL Number MFCD00054629
InChI Key VRAAQIWOSHYDHE-UHFFFAOYSA-M
Synonym 2-chlorobenzyl triphenylphosphonium chloride hydrate, 2-chlorophenyl methyl triphenylphosphanium hydrate chloride, acmc-20appm, 2-chlorobenzyl triphenylphosphoniumchloride
PubChem CID 60161216
IUPAC Name [(2-chlorophenyl)methyl]triphenylphosphanium chloride
SMILES [Cl-].ClC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C
Color White
Quantity 25 g
Formula Weight 423.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (2-Chlorobenzyl)triphenylphosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.