CAS RN 18592-13-7
IUPAC Name:
6-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
6-(chloromethyl)-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol):
160.56
Molecular Formula:
C5H5ClN2O2
InChi Key:
VCFXBAPEXBTNEA-UHFFFAOYSA-N
SMILES:
ClCC1=CC(=O)NC(=O)N1
1
–
2
of
2
results